6-Fluoro-3-methoxy-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

21663-1G 21663-500MG 21663-5G
76937-966EA 534.7 USD
76937-966 76937-968 76937-970
6-Fluoro-3-methoxy-2-methylpyridine ≥97%
6-Fluoro-3-methoxy-2-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611223
CAS Number: 1227581-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR