2-Fluoro-5-(hydroxymethyl)-6-picoline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (6-Fluoro-2-methylpyridin-3-yl)methanol, 6-Fluoro-3-hydroxymethyl-2-methylpyridine

21661-1G 21661-25G 21661-5G
76937-952EA 74.27 USD
76937-952 76937-954 76937-956
2-Fluoro-5-(hydroxymethyl)-6-picoline ≥97%
2-Fluoro-5-(hydroxymethyl)-6-picoline
Formula: C₇H₈FNO
MW: 141.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662401
CAS Number: 884494-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR