7-Bromo-4-chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

21534-1G 21534-25G 21534-500MG 21534-5G
76937-908EA 350.1 USD
76937-908 76937-910 76937-912 76937-914
7-Bromo-4-chloro-1H-indazole ≥97%
7-Bromo-4-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD09749923
CAS Number: 1000341-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR