5-Isopropoxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Isopropoxy-2-methylphenyl)boronic acid

21523-1G 21523-25G 21523-500MG 21523-5G
76937-892EA 157.02 USD
76937-892 76937-894 76937-896 76937-898
5-Isopropoxy-2-methylphenylboronic acid ≥97%
5-Isopropoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD19105377
CAS Number: 1451390-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR