5-Methoxynicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

21449-25G 21449-5G
76937-876EA 368.14 USD
76937-876 76937-878
5-Methoxynicotinic acid ≥97% research grade
5-Methoxynicotinic acid
Formula: C7H7NO3
Storage Temperature: Ambient
MDL Number: MFCD01603648
CAS Number: 20826-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR