2-Isopropoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21307-1G 21307-25G 21307-500MG 21307-5G
76937-846EA 171.87 USD
76937-846 76937-848 76937-850 76937-852
2-Isopropoxy-5-methylbenzaldehyde ≥97%
2-Isopropoxy-5-methylbenzaldehyde
Formula: C₁₁H₁₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18331386
CAS Number: 610797-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR