6-Bromo-5-fluoro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

21114-1G 21114-25G 21114-5G
76937-802EA 76.39 USD
76937-802 76937-804 76937-806
6-Bromo-5-fluoro-1H-indazole ≥97%
6-Bromo-5-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18089816
CAS Number: 1286734-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR