2-Fluoro-6-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoro-2-Iodobenzaldehyde

20989-25G 20989-5G
76937-782EA 407.39 USD
76937-782 76937-784
2-Fluoro-6-iodobenzaldehyde ≥97%
2-Fluoro-6-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FIO
MW: 250.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371506
CAS Number: 146137-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR