Methyl 6-chloro-2-fluoronicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2-fluoro-nicotinic acid methyl ester

20677-1G 20677-25G 20677-500MG 20677-5G
76937-760EA 245.07 USD
76937-760 76937-762 76937-764 76937-766
Methyl 6-chloro-2-fluoronicotinate ≥97% research grade
Methyl 6-chloro-2-fluoronicotinate
Formula: C₇H₅ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD16658093
CAS Number: 1093880-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR