6-Chloro-2-fluoro-3-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

20674-1G 20674-25G 20674-500MG 20674-5G
76937-746EA 441.34 USD
76937-746 76937-748 76937-750 76937-752
6-Chloro-2-fluoro-3-methoxypyridine ≥95%
6-Chloro-2-fluoro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610386
CAS Number: 1227573-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR