Ethyl 3,5-dibromo-4-pyridinecarboxylate ≥97%

Supplier: AOBChem USA

Warning

20629-1G 20629-25G 20629-500MG 20629-5G
76937-720EA 124.13 USD
76937-720 76937-722 76937-724 76937-726
Ethyl 3,5-dibromo-4-pyridinecarboxylate ≥97%
Ethyl 3,5-dibromo-4-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₇Br₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698215
CAS Number: 1214375-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR