Ethyl 2-bromo-6-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

20598-25G 20598-5G
76937-708EA 488.02 USD
76937-708 76937-710
Ethyl 2-bromo-6-chlorobenzoate ≥97%
Ethyl 2-bromo-6-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD07780717
CAS Number: 933672-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR