Methyl 2-methoxypyrimidine-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

20564-1G 20564-500MG 20564-5G
76937-698EA 593.05 USD
76937-698 76937-700 76937-702
Methyl 2-methoxypyrimidine-5-carboxylate ≥97%
Methyl 2-methoxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16660502
CAS Number: 38373-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR