Methyl 2,5-dibromo-3-pyridinecarboxylate ≥97%

Supplier: AOBChem USA

Warning

20559-1G 20559-25G 20559-5G
76937-686EA 76.39 USD
76937-686 76937-688 76937-690
Methyl 2,5-dibromo-3-pyridinecarboxylate ≥97%
Methyl 2,5-dibromo-3-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MW: 294.928 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698023
CAS Number: 78686-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR