Methyl-2-fluoro-5-(trifluoromethyl)benzenecarboxylate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Mthyl 2-fluoro-5-(trifluoromethyl)benzoate, Methyl 2-fluoro-5-(trifluoromethyl)benzoate

20483-25G 20483-5G
76937-674EA 204.76 USD
76937-674 76937-676
Methyl-2-fluoro-5-(trifluoromethyl)benzenecarboxylate ≥95%
Methyl-2-fluoro-5-(trifluoromethyl)benzenecarboxylate
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203706
CAS Number: 556112-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR