6-Bromo-3-chloropyridine-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

20417-1G 20417-25G 20417-5G
76937-660EA 92.3 USD
76937-660 76937-662 76937-664
6-Bromo-3-chloropyridine-2-carboxylic acid ≥97%
6-Bromo-3-chloropyridine-2-carboxylic acid
Formula: C₆H₃BrClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13185795
CAS Number: 1060815-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR