6-Bromo-3-methoxy-2-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

20397-1G 20397-25G 20397-500MG 20397-5G
76937-648EA 135.8 USD
76937-648 76937-650 76937-652 76937-654
6-Bromo-3-methoxy-2-methylpyridine ≥97% research grade
6-Bromo-3-methoxy-2-methylpyridine
Formula: C₇H₈BrNO
MW: 202.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846419
CAS Number: 850882-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR