tert-Butyl 4-chloro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

20322-1G 20322-25G 20322-5G
76937-622EA 62.6 USD
76937-622 76937-624 76937-626
tert-Butyl 4-chloro-3-methylbenzoate ≥97%
tert-Butyl 4-chloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₂
MW: 226.699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26378157
CAS Number: 1876229-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR