5-Tert-Butyl-2-ethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20148-1G 20148-25G 20148-5G
76937-598EA 96.55 USD
76937-598 76937-600 76937-602
5-Tert-Butyl-2-ethoxyphenylboronic acid ≥97%
5-Tert-Butyl-2-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD13195648
CAS Number: 1217501-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR