Methyl-5-bromo-2-ethoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-Bromo-2-ethoxynicotinate

20120-1G 20120-25G 20120-5G
76937-590EA 100.79 USD
76937-590 76937-592 76937-594
Methyl-5-bromo-2-ethoxynicotinate ≥97%
Methyl-5-bromo-2-ethoxynicotinate
Formula: C₉H₁₀BrNO₃
MW: 260.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09842613
CAS Number: 1031927-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR