5-Iodo-2-isopropoxypyridine ≥95%

Supplier: AOBChem USA

Warning

20059-1G 20059-25G 20059-500MG 20059-5G
76937-574EA 173.99 USD
76937-574 76937-576 76937-578 76937-580
5-Iodo-2-isopropoxypyridine ≥95%
5-Iodo-2-isopropoxypyridine
Formula: C₈H₁₀INO
Storage Temperature: Refrigerator
MDL Number: MFCD07781185
CAS Number: 902837-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR