3-Bromophenyl Diethylcarbamate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Carbamic acid, N,N-diethyl-, 3-bromophenyl ester

19926-1G 19926-25G 19926-5G
76937-552EA 62.6 USD
76937-552 76937-554 76937-556
3-Bromophenyl Diethylcarbamate ≥97%
3-Bromophenyl Diethylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₂
MW: 272.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27930373
CAS Number: 863870-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR