7-Fluoro-1H-indazole-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 7-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

19882-1G 19882-25G 19882-500MG 19882-5G
76937-536EA 260.99 USD
76937-536 76937-538 76937-540 76937-542
7-Fluoro-1H-indazole-4-boronic acid pinacol ester ≥97%
7-Fluoro-1H-indazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD19687061
CAS Number: 1186334-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR