Methyl-5-bromo-2-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-bromo-2-methoxynicotinate, Methyl 5-bromo-2-methoxy-3-pyridinecarboxylate

19776-25G 19776-5G
76937-514EA 251.44 USD
76937-514 76937-516
Methyl-5-bromo-2-methoxynicotinate ≥97%
Methyl-5-bromo-2-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD08064227
CAS Number: 122433-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR