3-Iodo-2-(2,2,2-trimethylacetamido)pyridine ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: N-(3-Iodopyridin-2-yl)pivalamide, N-(3-Iod-2-pyridyl)pivalamide, N-(3-Iodopyridin-2-yl)-2,2-dimethylpropionamide

19638-25G 19638-5G
76937-490EA 262.05 USD
76937-490 76937-492
3-Iodo-2-(2,2,2-trimethylacetamido)pyridine ≥95% research grade
N-(3-Iod-2-pyridyl)pivalamide
Formula: C₁₀H₁₃IN₂O
MW: 304.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662384
CAS Number: 113975-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR