N-(4-Bromo-5-fluoro-2-methylphenyl)acetamide ≥97%
Supplier: AOBChem USA
|
Warning
|
19342-25G
19342-5G
76937-468EA
524.09
USD
76937-468
76937-470
N-(4-Bromo-5-fluoro-2-methylphenyl)acetamide ≥97%
N-(4-Bromo-5-fluoro-2-methylphenyl)acetamide
Precaución: for Research Use Only
Formula:
C₉H₉BrFNO Storage Temperature: Ambient |
MDL Number:
MFCD22989047 CAS Number: 633335-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...