2-Ethoxy-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Benzoic acid, 2-ethoxy-5-formyl-

19147-1G 19147-25G 19147-500MG 19147-5G
76937-438EA 138.98 USD
76937-438 76937-440 76937-442 76937-444
2-Ethoxy-5-formylbenzoic acid ≥97%
2-Ethoxy-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05625188
CAS Number: 848606-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR