tert-Butyl 3-bromo-4-chlorophenylcarbamate ≥97%

Supplier: AOBChem USA

Warning

19116-1G 19116-25G 19116-500MG 19116-5G
76937-422EA 137.92 USD
76937-422 76937-424 76937-426 76937-428
tert-Butyl 3-bromo-4-chlorophenylcarbamate ≥97%
tert-Butyl 3-bromo-4-chlorophenylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrClNO₂
MW: 306.583 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23832100
CAS Number: 1080573-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR