(6-(Pyrrolidin-1-yl)pyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

18835-1G 18835-25G 18835-5G
76937-392EA 73.21 USD
76937-392 76937-394 76937-396
(6-(Pyrrolidin-1-yl)pyridin-3-yl)boronic acid ≥97%
(6-(Pyrrolidin-1-yl)pyridin-3-yl)boronic acid
Formula: C₉H₁₃BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06801704
CAS Number: 1150114-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR