tert-Butyl 4-bromo-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

18714-1G 18714-25G 18714-500MG 18714-5G
76937-352EA 124.13 USD
76937-352 76937-354 76937-356 76937-358
tert-Butyl 4-bromo-2-methylbenzoate ≥95%
tert-Butyl 4-bromo-2-methylbenzoate
Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08703427
CAS Number: 445003-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR