tert-Butyl 4-bromo-2,6-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

18710-25G 18710-5G
76937-342EA 357.53 USD
76937-342 76937-344
tert-Butyl 4-bromo-2,6-difluorobenzoate ≥97%
tert-Butyl 4-bromo-2,6-difluorobenzoate
Formula: C₁₁H₁₁BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08703422
CAS Number: 955887-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR