6-Methoxy-2-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Methoxy-2-methylpyridine-3-carboxaldehyde

18516-1G 18516-25G 18516-500MG 18516-5G
76937-330EA 195.21 USD
76937-330 76937-332 76937-334 76937-336
6-Methoxy-2-methylnicotinaldehyde ≥97%
6-Methoxy-2-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044370
CAS Number: 156094-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR