N-(3-Chlorophenyl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-(3-Chloro-phenyl)-2,2-dimethyl-propionamide

18337-1G 18337-25G 18337-500MG 18337-5G
76937-304EA 138.98 USD
76937-304 76937-306 76937-308 76937-310
N-(3-Chlorophenyl)pivalamide ≥97%
N-(3-Chlorophenyl)pivalamide
Formula: C₁₁H₁₄ClNO
MW: 211.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02310087
CAS Number: 32597-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR