6-Bromo-2-chloro-3-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

18192-1G 18192-25G 18192-5G
76937-276EA 90.18 USD
76937-276 76937-278 76937-280
6-Bromo-2-chloro-3-ethoxybenzoic acid ≥97%
6-Bromo-2-chloro-3-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrClO₃
MW: 279.515 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631775
CAS Number: 2379321-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR