(5-Phenylpyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

18099-1G 18099-25G 18099-500MG 18099-5G
76937-264EA 171.87 USD
76937-264 76937-266 76937-268 76937-270
(5-Phenylpyridin-3-yl)boronic acid ≥97%
(5-Phenylpyridin-3-yl)boronic acid
Formula: C₁₁H₁₀BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08061934
CAS Number: 850991-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR