7-Bromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

17859-25G 17859-5G
76937-246EA 233.4 USD
76937-246 76937-248
7-Bromo-1H-indazole ≥97%
7-Bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD07371564
CAS Number: 53857-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR