6-Bromo-3-methyl-1H-indazole ≥95%
Supplier: AOBChem USA
|
Warning
|
17844-25G
17844-5G
76937-234EA
371.32
USD
76937-234
76937-236
6-Bromo-3-methyl-1H-indazole ≥95%
6-Bromo-3-methyl-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08272244 CAS Number: 7746-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...