6-Methoxy-5-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

17723-1G 17723-25G 17723-500MG 17723-5G
76937-222EA 260.99 USD
76937-222 76937-224 76937-226 76937-228
6-Methoxy-5-methylnicotinaldehyde ≥97%
6-Methoxy-5-methylnicotinaldehyde
Formula: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD11856047
CAS Number: 123506-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR