6-Fluoro-N,N-dimethylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

17498-1G 17498-25G 17498-500MG 17498-5G
76937-202EA 123.07 USD
76937-202 76937-204 76937-206 76937-208
6-Fluoro-N,N-dimethylpyridin-2-amine ≥97%
6-Fluoro-N,N-dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₉FN₂
MW: 140.158 g/mol
Storage Temperature: Ambient
CAS Number: 909187-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR