6-Benzyloxy-2-fluoro-3-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17407-1G 17407-25G 17407-5G
76937-192EA 109.28 USD
76937-192 76937-194 76937-196
6-Benzyloxy-2-fluoro-3-methylphenylboronic acid ≥97%
6-Benzyloxy-2-fluoro-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄BFO₃
MW: 260.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856037
CAS Number: 1451391-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR