Methyl 4-acetyl-3-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

16741-1G 16741-500MG 16741-5G
76937-152EA 471.04 USD
76937-152 76937-154 76937-156
Methyl 4-acetyl-3-fluorobenzoate ≥95%
Methyl 4-acetyl-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
MW: 196.175 g/mol
Storage Temperature: Ambient
CAS Number: 1059549-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR