(3-(Methoxycarbonyl)-4-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Benzoic acid, 5-borono-2-methyl-, 1-methyl ester

16439-1G 16439-25G 16439-500MG 16439-5G
76937-124EA 118.83 USD
76937-124 76937-126 76937-128 76937-130
(3-(Methoxycarbonyl)-4-methylphenyl)boronic acid ≥97% research grade
(3-(Methoxycarbonyl)-4-methylphenyl)boronic acid
Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13191800
CAS Number: 1048330-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR