6-Bromo-2,3,4-trifluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16295-1G 16295-25G 16295-500MG 16295-5G
76937-098EA 124.13 USD
76937-098 76937-100 76937-102 76937-104
6-Bromo-2,3,4-trifluorobenzaldehyde ≥95%
6-Bromo-2,3,4-trifluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₂BrF₃O
MW: 238.989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527216
CAS Number: 372519-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR