6-Bromo-2-fluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16286-1G 16286-25G 16286-500MG 16286-5G
76937-082EA 201.58 USD
76937-082 76937-084 76937-086 76937-088
6-Bromo-2-fluoro-3-hydroxybenzaldehyde ≥97%
6-Bromo-2-fluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD09261253
CAS Number: 935534-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR