6-Bromo-2,3-difluorophenol ≥97% research grade

Supplier: AOBChem USA

Warning

16033-1G 16033-25G 16033-5G
76937-064EA 88.06 USD
76937-064 76937-066 76937-068
6-Bromo-2,3-difluorophenol ≥97% research grade
6-Bromo-2,3-difluorophenol
Formula: C₆H₃BrF₂O
MW: 208.988 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11656842
CAS Number: 186590-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR