6-Bromo-2,3-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15702-1G 15702-25G 15702-500MG 15702-5G
76937-034EA 231.28 USD
76937-034 76937-036 76937-038 76937-040
6-Bromo-2,3-dichlorobenzaldehyde ≥97%
6-Bromo-2,3-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O
MW: 253.908 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526367
CAS Number: 945999-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR