6-Bromo-2-fluoro-3-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15615-1G 15615-25G 15615-500MG 15615-5G
76937-022EA 171.87 USD
76937-022 76937-024 76937-026 76937-028
6-Bromo-2-fluoro-3-hydroxyphenylboronic acid ≥97%
6-Bromo-2-fluoro-3-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD12026728
CAS Number: 1309980-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR