6-Bromo-3-chloro-2-fluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
15605-1G
15605-25G
15605-5G
76937-010EA
117.76
USD
76937-010
76937-012
76937-014
6-Bromo-3-chloro-2-fluorobenzaldehyde ≥97%
6-Bromo-3-chloro-2-fluorobenzaldehyde
Formula:
C₇H₃BrClFO MW: 237.454 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11520661 CAS Number: 1114809-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...