6-Bromo-2-fluoro-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15602-1G 15602-25G 15602-500MG 15602-5G
76936-996EA 385.11 USD
76936-996 76936-998 76937-000 76937-002
6-Bromo-2-fluoro-3-formylphenylboronic acid ≥97%
6-Bromo-2-fluoro-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrFO₃
MW: 246.826 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026727
CAS Number: 1315340-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR