tert-Butyl-4-(trifluoromethoxy)phenylcarbamate ≥97%

Supplier: AOBChem USA

Warning

15521-1G 15521-25G 15521-500MG 15521-5G
76936-980EA 162.32 USD
76936-980 76936-982 76936-984 76936-986
tert-Butyl-4-(trifluoromethoxy)phenylcarbamate ≥97%
tert-Butyl-4-(trifluoromethoxy)phenylcarbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD09910418
CAS Number: 212696-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR